Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stanbio™ Glycohemoglobin (HbA1c) Bi-Level Control
SDP

For use in clinical testing

Supplier:  Stanbio™ 0357201

Catalog No. SB-0357-201

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Description

Description

  • Volume: 2 × 1mL

Specifications

Specifications

-20°C
Clinical testing
2 x 1 mL
Glycohemoglobin Control
2
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit