Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stanbio™ BHB Tri Level Control (6x5mL/Ea)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Designed for maximum stability and ease-of-use
Supplier: Stanbio™ 2460605
This item is not returnable.
View return policy
Description
- Enzymatic beta-hydroxybutyrate reaction
- Ready-to-use liquid reagents
- Use serum or plasma
- Linear up to 4.5 mmol/L
- Read at 505 nm
Specifications
CE Marked | |
Quantitative testing | |
Liquid |
BHB Tri Level Control | |
6 x 5 mL |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction