Learn More
Wilmad Labglass STALAGMOMETER TUBE ONLY 5.0ML
Supplier: Wilmad Labglass LG5050106
Stalagmometer, Tube Only. Size: 5mL. Specifically designed for checking the surface tension of liquids by drop count method.