Learn More
Wilmad Labglass STALAGMOMETER TUBE ONLY 2.5 ML
Supplier: Wilmad Labglass LG5050102
Stalagmometer, Tube Only. Size: 2.5mL. Specifically designed for checking the surface tension of liquids by drop count method.