Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Stak-a-Cab™ Stackable Cabinets
Use individually or stack up to three high with the optional floor stand.
Supplier: SciMatCo SC1860/EMD
Includes: Each part number represent one side-by-side cabinet; set of three cabinets shown represents three separate items stacked with base.
Description
No interior metal parts to corrode: wooden hinges and handles, polypropylene lining on cabinet floor, and polypropylene padlock assembly.
- Lock with two keys and piano-style door hinges
- All hardware is corrosion resistant
- Liquid-tight trough floors
- 10 x 10 in. (25 x 25cm) Hazard labels on both doors
- Epoxy paint finish inside and out
Corrosives Cabinet
- Capacity: 22 × 2.5L (approx. 14.5 gal./55L)
- Blue finish
- Accepts nitric acid compartment
- Capacity: 16 × 1 gal. (approx. 60.5L)
- Yellow finish
- Combination corrosives/flammables cabinet
- Corrosives compartment capacity: 10 × 2.5L (approx. 6.5 gal./25L)
- Flammables compartment capacity: 8 × 1 gal. (approx. 30L)
- Gray finish
- Height: 4.5 in. (11cm)
- Epoxy paint finish
Order Info
Interior: 15.125D x 45W x 16 in.H (40 x 114 x 41cm); Exterior: 18L x 47W x 18 in.H (46 x 119 x 46cm)
Compliance
Meet all applicable OSHA and NFPA standards.
Specifications
2 | |
119.4 cm | |
46 cm | |
18 in. | |
NFPA, OSHA | |
Storage Cabinet | |
16 x 1 gal. | |
Wood (Plywood) |
Epoxy | |
18 in. | |
Flammables | |
18 x 47 x 18 in. (46 x 119.4 x 45.7 cm) | |
47 in. | |
Manual Close | |
Yellow |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction