Learn More
Andwin Scientific STAINLESS STEEL RACK FOR MULTI
Supplier: Andwin Scientific 26420957
The 8-section (4 x 2) upright freezer rack is able to store 56 standard size 96-well plates with lids, 64 without lids or any plate or rack with a 5" x 3 3/8" or smaller footprint. This product is resistant against corrosion and have solid sides with handles. The back panels are also accessible to permit easy removal of plates by simply being pushed forward. The rack measures 14 3/4" L x 5 1/2" W x 9 1/4" H.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.