Learn More
United Scientific Supplies Stainless Steel Mixing Bowls
SDPSCI_ED
Mixing bowls are constructed from 304 stainless steel for increased heat resistance.
Supplier:  United Scientific Supplies BMX800
 View more versions of this product
Catalog No. S43769
Add to Cart
Description
Description
  • United Mixing Bowls are constructed from 304 stainless steel for increased heat resistance.
  • Bowls feature rounded corners, and polished mirror surface for easy mixing and cleaning.
  • Can withstand temperatures up to 1472°F (800°C).
  • Meets NSF standards.
Specifications
Specifications
Stainless Steel
13.25 in.
Round
8 qt.
5.38 in.
Mixing Bowl
Product Suggestions
Product Suggestions