Learn More
Stainless Steel gauze, 80 mesh woven from 0.127mm (0.005in) dia wire, Type 304, Thermo Scientific Chemicals
$53.65 - $230.65
Specifications
Chemical Name or Material | Stainless Steel gauze, Type 304 |
---|---|
Name Note | Woven from 0.127mm (0.005 in.) dia. wire |
Avg. Mol. Wt. or Mol. Wt. Range | Fe:Cr:Ni; 70:19:11 wt% |
MDL Number | MFCD00148499 |
Solubility Information | Insoluble in water. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA13477RR
|
Thermo Scientific Chemicals
013477RR |
10 x 10 cm |
Each
for $53.65
Save 12% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA13477CH
|
Thermo Scientific Chemicals
013477CH |
20 x 30 cm |
Each
for $107.65
Save 12% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
AA13477RU
|
Thermo Scientific Chemicals
013477RU |
30 x 60 cm |
Each
for $230.65
Save 13% Reg :
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
It is most commonly utilized for wire cloth weaving. Typical wire cloth applications include use with chemicals, food products, pharmaceuticals and exposure to moisture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel gauze, Type 304 | |
Fe:Cr:Ni; 70:19:11 wt% | |
Insoluble in water. | |
Wire Cloth |
Woven from 0.127mm (0.005 in.) dia. wire | |
MFCD00148499 | |
80 mesh |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.