Learn More
Stainless Steel gauze, 20 mesh woven from 0.382mm (0.015in) dia wire, Type 304, Thermo Scientific Chemicals
Filtration, electrodes
Supplier: Thermo Scientific Chemicals 040942GT
Description
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Stainless Steel gauze, Type 304 | |
Fe:Cr:Ni; 70:19:11 wt% | |
150 x 150 mm | |
20 mesh |
Woven from 0.382mm (0.015 in.) dia. wire | |
MFCD00148499 | |
Insoluble in water. | |
Wire Cloth |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
RUO – Research Use Only