Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Berkshire™ Stainless Steel Flat Mop Sieve

For efficient cleaning of floors, walls, ceilings and other large surfaces. Berkshire Stainless Steel Flat Mop Sieve is made of high quality stainless steel and is fully autoclavable.
Supplier: Berkshire™ BCRFMSIEVE1
Description
- Made of high quality stainless steel
- For one-step wet cleaning process
- 48.2cm (19 in.) wide
Specifications
Autoclavable | |
Silver | |
1/Pk. |
Stainless Steel | |
19 in. | |
Flat Mop Sieve |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title