Learn More
Troemner™ Stainless Steel Class 7 Weight Set
Manufactured from stainless steel. The Troemner™ Stainless Steel Class 7 Weight Set is used for rough weighing operations in physical and chemical laboratories.
Supplier: Troemner™ 30390970
Description
- One- and two-piece construction, depending on weight
- Includes certificate of accuracy
- Includes case with latch
- Made in the USA
Specifications
Weight Set with No Certificate | |
Stainless Steel | |
50g, (2) 20g, 10g, 5g, (2) 2g, 1g, 500mg, (2) 200mg, 100mg, 50mg, (2) 20mg, 10mg |
Class 7 | |
Plastic |