Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Stainless Steel Class 1 Cleanroom Lab Table - PROMO
SDP

With LisStat™ ESD Laminate and rounded front edge. Table features an adjustable height range with 2-inch increments and an easy 3 minute assembly

 View more versions of this product

Catalog No. 25-100-492PM

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

  • Grade 304 solid stainless steel frame and white sheet top
  • Not perforated
  • No.4 brushed finish stainless
  • Easy to clean
  • Meets and exceeds SEFA
  • Work Surface: ESD Laminate; static dissipative laminate sheets
Specifications

Specifications

Cleanroom Table
1524 mm
762 to 914 mm
Bench Frame, legs and top
5,000 lb
30 to 36 in. (2 in. increments)
762 mm
ANSI / BIFMA, SEFA
60 in.
25 year unconditional manufacturer warranty
2 in.
Stainless
30 to 36 in.
30 in.
White
Stainless Steel (304)
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit