Learn More
Fine Science Tools Burrs for Micro Drill, Stainless Steel, 1.4 mm Tip Dia, 44 mm, 10/pk
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Fine Science Tools 1900814
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Tip Diameter: 1.4mm/ Shaft Diameter: 2.3mm/ Alloy / Material: Stainless Steel/ Length: 44mm/ Autoclave Safe: Yes