Learn More
Azzota Corp Stage Micrometers Slide (Stage Graticules) ; Ideal For Microscope Calibration And Shading Correction. Vertical, Horizontal And Diameter Calibration. Accuracy: <=3% ; Verifying Aspect Ratio
Supplier: Azzota Corp TSM2
Stage Micrometers Slide (Stage Graticules) ; Ideal for microscope calibration and shading correction. Vertical, horizontal and Diameter calibration. Accuracy: <=3% ; verifying aspect ratio