Learn More
World Precision Instrument Stage Micrometer, 3 Ranges
Supplier: World Precision Instrument 504606
- 1X3" slide with linear targets etched into the glass
- 3 ranges: 10 mm / 100 to 0.1 mm (100 um) scale, 10 mm / 200 to 0.05 mm (50 um) scale and 1 mm / 100 to 0.01 mm (10μm) small scale