Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Stackable Low-Profile Lowboys™ HDPE Carboy
Click to view available options
Capacity (Metric):
8 L
15 L
Description
- Flat, rectangular design permits storing in limited space
- Made of durable high-density chemically resistant polyethylene
- Polypropylene (PP) spigot and leakproof (PP) closure
- Special contour handle allows easy, convenient and integral one-hand carrying even when full
- Supplied with four acid-proof labels
Specifications
Product Type | Low-profile Carboy |
Capacity (English) | 4 gal. |
Closure Material | Polypropylene |
Closure Size | 63mm |
Capacity (Metric) | 15 L |
Material | HDPE |
For Use With (Application) | Dispense most liquids, including reagents, distilled water and titrants. |
Includes | Four Acid-proof labels |
Length (Metric) | 371 mm |
Color | White |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction