Learn More
Hach Company Stablcal Reference Suspension IV, 30 NTU, 100 mL
Supplier: Hach Company 2897042
Stabilised Formazine reference suspension for the determination of lucidity/opalescence according to European Pharmacopeia 6.0.