Learn More
Specialty Silicone Products ST2060 .125" (3mm) 100/pack
Supplier:  Specialty Silicone Products 07002007C
Encompass_Preferred
ST2060 .125" (3mm) Thick, Natural Ptfe/Clear Silicone, Ultra Low Bleed, Epa Septa Inserted Into 20Mm Silver Aluminum Crimp Cap Pk/100. Must order minimum of 6 packs.
Catalog No. NC2004803