Learn More
Across International SST Storage Drawers for Ai G20h -86C Freezers 40,000 Vials Max.
SDP
Supplier:  Across International DRAWERSETG20H
Encompass_Preferred
Stainless steel drawer sets for Ai RapidChill -86C ultra-low freezers. Price includes 16 drawer sets and 80 drawers. This listing is for the stainless steel storage drawers ONLY, vials, vial boxes and ULT freezer sold separately.
Catalog No. 50-225-1373