Learn More
Bellco Glass, Inc. 316L stainless steel port assemblies are designed for use with media storage bottles for adding and withdrawing media. 3-port models feature siphon, fill and vent tubes and is supplied with a 45mm polypropylene blue open top screw cap and white silicone g
SDP
Supplier:  Bellco Glass, Inc. 563730010R
Encompass_Preferred
316L stainless steel port assemblies are designed for use with media storage bottles for adding and withdrawing media. 3-port models feature siphon, fill and vent tubes and is supplied with a 45mm polypropylene blue open top screw cap and white silicone g
Catalog No. 50-194-5784