Learn More
Wesco Industrial Products 304 Stainless Steel 55 Gallon Drum Dolly
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Wesco Industrial Products 240197
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
55 Gallon 304 Stainless Steel Drum Dolly. Casters have a 304 stainless steel rig with 3" white polypropylene wheel and stainless steel hardware. Suitable for wash down. Stability Advantage - Casters are mounted past the dolly and drum rim. Our casters are mounted further apart than standard designs. The further the casters are apart, the greater the stability. 24" ID accommodates a standard 55 gallon drum.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.