Learn More
Ace Glass, Inc. Stainless steel support rod 61cm 24"
Supplier: Ace Glass, Inc. 1117808
24 (61cm) long, 1/2 (13mm) O.D. 303 stainless steel support rods, for use in place of aluminum or fiberglass rods. Can be used with all standard laboratory connectors.