Learn More
Chemglass Life Sciences Dish Only, 200 x 100mm, Oil Bath, Stainless Steel
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG1100D03
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Dish Only, 200 x 100mm, Oil Bath, Stainless Steel. Stainless steel dish with lid for use as an oil bath when used in conjunction with a hot plate stirrer having a thermocouple connection. Available in three sizes, complete with lids.