Learn More
Andwin Scientific SS 30-B. SUBTILIS NCTC 10400
SDP
Supplier:  Andwin Scientific 95060888
Encompass_Preferred
BioBall SS 30-B. SUBTILIS NCTC 10400SingleShot delivers 30 cfu with no prep time! Offers unparalleled precision ease of use and is ideal for food and pharmaceutical applications requiring an ultra-low known level of inoculum
Catalog No. NC2002799
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by Fisher Scientific.