Learn More
MilliporeSigma™ Supelco™ SS-174 PTFE-Faced Septa
Suitable to overcome the degradation of the septa, due to prolonged high temperatures which occurs in silicone rubber septa
Supplier:  MilliporeSigma™ Supelco™ 22732
 View more versions of this product
Catalog No. 11-101-0718
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
  • Septa degradation results in baseline drift and additional peaks
  • When the PTFE is faced towards the column packing this issue is eliminated
  • Designed for applications where greater inertness is required
  • The off-white silicone rubber body (123 mil) provides plenty of support for the yellow PTFE-face (2 mil)
  • After the PTFE is ruptured during injection, some bleeding may occur
  • It is recommend to use a needle guide to minimize the size of the punctured area
  • For use with 200°C to 300 °C inlet temperatures
Specifications
Specifications
PTFE Face
14 mm
9/16 in.
Silicone
123 mil
300°C
SS-174 Septa
50 Pk.
Product Suggestions
Product Suggestions