Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SR 0987
T cell-specific RORγ (RORγt) agonist
$242.44 - $1010.36
Specifications
Chemical Name or Material | 2-Chloro-N-[4-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl]phenyl]benzamide |
---|---|
CAS | 303126-97-8 |
Target | Nuclear Receptor Agonists |
Molecular Formula | C16H10ClF6NO2 |
Formula Weight | 397.7 |
Description
T cell-specific RORγ (RORγt) agonist (EC50 = 800 nM in a gene reporter assay). Induces IL-17 expression and decreases PD-1 expression in T cells in vitro.Specifications
2-Chloro-N-[4-[2,2,2-trifluoro-1-hydroxy-1-(trifluoromethyl)ethyl]phenyl]benzamide | |
Nuclear Receptor Agonists | |
397.7 |
303126-97-8 | |
C16H10ClF6NO2 | |
>98% |
Safety and Handling
Recommended Storage : Store at 4°C
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title