Learn More
Invivogen AddaVax™, Squalene-oil-in-water, 10mL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Invivogen VACADX10
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
Specificity: Th1/Th2 response. Working concentration: 1:1 (AddaVax™:antigen). Quality control: Sterility guaranteed, Endotoxin level < 1 EU/ml, Particle size ~ 160 nm. AddaVax™ is a squalene-based oil-in-water nano-emulsion.