Learn More
Cole-Parmer Argos Technologies Cell Spreaders, Sterile, Polypropylene, 50 bags of 10; 500/Cs
Supplier: Cole-Parmer 0439651
Spread bacterial cultures evenly with these cell spreaders. Apply cultures over an entire petri dish or plate with the L-shaped spreaders, while reducing the risk of damaging the agar. They have a smooth spreading surface with slight upward return.