Learn More
PERFORMANCE HEALTH Quick Freeze Cold Spray
Supplier:  PERFORMANCE HEALTH 081565464
Encompass_Preferred
Quick Freeze Cold Spray speeds the cooling of low-temperature thermoplastics without leaving any residue. It can also be used on rubbers and metals. The Cold Spray is odorless, non-flammable and should not be used on skin.
Catalog No. NC1691810