Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SPP 86
Potent RET inhibitor
$327.50 - $1367.00
Specifications
Purity | >98% |
---|---|
Description | SPP 86 |
Molecular Weight (g/mol) | 277.32 |
Product Type | SPP 86 |
Description
Potent RET inhibitor (IC50 = 8 nM). Also exhibits inhibitory activity at EphA1, FGFR1, Flt4, Lck and Yes. Inhibits proliferation of HCT 116 and TPC1 cells in vitro. Cell permeable.Specifications
>98% | |
SPP 86 | |
277.32 | |
SPP 86 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title