Learn More
VWR INTERNATIONAL INC Fine-Pore Cellulose Sponges, 14 x 9.2 x 2.5 cm, 12/pk
Supplier: VWR INTERNATIONAL INC 58540047
These fine pore natural color cellulose sponges hold about 25 times their own weight in water. Resist tearing and abrasion. Absorb liquid more evenly than large pore sponges. Available in compressed (approximately 1/8" thickness before expansion) or uncompressed format.