Learn More
Sklar Instruments Sponge Bowl, Sklar, Stainless Steel. This bowls measures 5 in. x 2-3/8 in. and has a capacity of 22 ounces. EA/1.
Supplier: Sklar Instruments 101498
Sponge Bowl, Sklar, Stainless Steel. Sklar Sponge Bowls are most commonly utilized as a recepticle for contaminated sponges to be placed throughout a surgical procedure. They may; however, also be used as a container for holding sponges or sponge sticks that have been soaked in a sterile solution to be used during a procedure. This bowls measures 5 in. x 2-3/8 in. and has a capacity of 22 ounces.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.