Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Supelco™ SPME Fiber Assembly, Carboxen/Polydimethylsiloxane (CAR/PDMS), df 85 μm
For Gases and Low Molecular Weight Compounds (MW 30-225)
$793.00 - $1031.00
Catalog Number | Mfr. No. | For Use With (Equipment) | Material | Needle Gauge | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Material | Needle Gauge | Price | Quantity & Availability | |||
11-100-7790
|
MilliporeSigma™ Supelco™
57906U |
Autosampler | Metal Alloy Fiber | 23 ga |
Each for $1,031.00
|
|
|||
11-102-0743
|
MilliporeSigma™ Supelco™
57295U |
Autosampler | StableFlex Fiber | 23 ga |
Each for $969.00
|
|
|||
11-100-8573
|
MilliporeSigma™ Supelco™
57335U |
Autosampler | StableFlex Fiber | 24 ga |
Each for $800.00
|
|
|||
11-100-8583
|
MilliporeSigma™ Supelco™
57334U |
Manual Holder | StableFlex Fiber | 24 ga |
Each for $793.00
|
|
|||
Description
Solid phase microextraction (SPME) technique is generally used for GC/MS analysis. It helps in simplifying and speeding up the sample preparation. It also reduces solvent requirement during extraction.Specifications
SPME | |
SPME Fibers & Accessories |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title