Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Splitomicin, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Inhibitor of Sir2p (IC50 = 60 μM), an NAD+-dependent Sir2 family deacetylase required for chromatin-dependent silencing in yeast.
Specifications
Quantity | 50 mg |
Synonym | splitomicin, 1,2-dihydro-3h-naphtho 2,1-b pyran-3-one, 1,2-dihydro-3h-benzo f chromen-3-one, 1h-benzo f chromen-3 2h-one, chembl86537, 1,2-dihydrobenzo f chromen-3-one, splitomycin, tocris-1542, d09suo, splitomicin hplc , powder |
SMILES | C1CC(=O)OC2=C1C3=CC=CC=C3C=C2 |
IUPAC Name | 1,2-dihydrobenzo[f]chromen-3-one |
PubChem CID | 5269 |
Formula Weight | 198.22 |
Percent Purity | >98% |
Chemical Name or Material | Splitomicin |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction