Learn More
Across International 1200C 1-Zone 50mm OD Top-Open Tube Furnace with Quartz Tube
SDP
Supplier:  Across International STF1200.50.600
Encompass_Preferred
STF1200 1200C Max. Split Tube Furnace with 50x600mm Quartz TubeNon UL approved
Catalog No. NC2156188