Learn More
Across International 1200C 1-Zone 50mm OD Top-Open Tube Furnace with Quartz Tube
SDP
Supplier:  Across International STF1200.50.600UL
Encompass_Preferred
STF1200 50x600mm (ODxlength) portable split tube furnaces feature spiral wire coils embedded in Mitsubishi (Japan) alumina fiber insulation. A double walled steel internal housing helps minimize heat loss to exterior surface. Furnace operation is controlled by a 30-segment digital controller. All our furnaces are CE compliant.
Catalog No. NC2155874