Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spironolactone, USP, 97-103%, Spectrum™ Chemical
SDP

Catalog Number 18606979
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
52-01-7
C24H32O4S
416.58
LXMSZDCAJNLERA-ZHYRCANASA-N
(1R,3aS,3bR,4R,9aR,9bS,11aS)-4-(acetylsulfanyl)-9a,11a-dimethyl-2,3,3a,3b,4,5,7,8,9,9a,9b,10,11,11a-tetradecahydrospiro[cyclopenta[a]phenanthrene-1,2'-oxolane]-5',7-dione
CC(=O)S[C@@H]1CC2=CC(=O)CC[C@]2(C)[C@H]2CC[C@@]3(C)[C@@H](CC[C@@]33CCC(=O)O3)[C@H]12
52-01-7
C24H32O4S
416.58
LXMSZDCAJNLERA-ZHYRCANASA-N
(1R,3aS,3bR,4R,9aR,9bS,11aS)-4-(acetylsulfanyl)-9a,11a-dimethyl-2,3,3a,3b,4,5,7,8,9,9a,9b,10,11,11a-tetradecahydrospiro[cyclopenta[a]phenanthrene-1,2'-oxolane]-5',7-dione
CC(=O)S[C@@H]1CC2=CC(=O)CC[C@]2(C)[C@H]2CC[C@@]3(C)[C@@H](CC[C@@]33CCC(=O)O3)[C@H]12

SP320, 52-01-7, C24H32O4S

Spectrum™ Chemical Spironolactone, USP is used to treat high blood pressure and edema. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

CAS Max % 1
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 100 g
Specific Rotation -41° to -45°
Percent Purity 97 to 103%
Grade USP
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit