Learn More
Andwin Scientific SPHER HTG MNTL 450F/500ML BKR
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Andwin Scientific 100BTM106
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Spherical flask mantle only covers the bottom half of the 500 mL flasks, allowing contents to be fully visible. Its exterior is made with durable aluminum, while its interior is constructed with fabric in order to provide soft surroundings for glass vessels. This product can resist 450C (840F) internal operating temperature. This is sold with detachable, 1.2m (4') long, three-wire cord and locking connector.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.