Learn More
Azzota Corp Calibration Filters Set, 8 Piece: 10%T Neutral Density, 20%T Neutral Density, 30%T Neutral Density, 220Nm Stray Light, 360Nm Stray Light, Holmium , Didymium , Blank
Supplier: Azzota Corp Q808
Calibration Filters Set, 8 piece calibration set: #1 - 10%T neutral density filter, #2 - 20%T neutral density filter, #3 - 30%T neutral density filter, #4 - 220nm stray light filter, #5 - 360nm stray light filter, #6 - Holmium filter, #7 - Didymium filter, #8 - BLANK filter, Accuracy: +/-0.2nm or +/-1%, mounted in a standard 10mm cuvette metal holder
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.