Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Honeywell™ Sperian™ Survivair Opti-Fit Air-Purifying Respirator Masks
Specifications
Color | Black |
---|---|
For Use With (Equipment) | Opti-fit™ air purifying respirator |
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
17-377-462
|
Honeywell™
702008 |
D-Ring |
Each of 1 for $1.98
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Specifications
Black | |
Opti-fit™ air purifying respirator |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title