Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Spectrum™ Spectra Mesh™ Woven, Nylon Filters
Consist of individual strands woven into a mesh screen
Supplier: Spectrum™ 148134
Description
Combines high tensile strength with flexibility
- Water Affinity: Hydrophillic
- Strength: High
- Adsorption: High
- Best Resistance: Corrosives
- pH Resistance: pH 3-10
- Thermal Stability: up to 180°C
- Sterilization: Irradiation
Specifications
Disc | |
20 μm | |
55 μm |
Nylon | |
Hazardous waste, drinking water purification, Acid rain measurement, suspended soils, air/asbestos, coal dust analysis, oceanography, trace element analysis | |
47 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction