Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Spectra™ Multicolor Broad Range Protein Ladder

Thermo Scientific™ Spectra Multicolor Broad Range Protein Ladder is a 4-color protein standard containing 10 prestained proteins (10 to 260 kDa) for use as size standards in gel electrophoresis and Western blotting.

Supplier:  Thermo Scientific™ 26634X4

Encompass_Preferred

 View more versions of this product

Catalog No. 26-634-X4


Add to Cart

Description

Description

Thermo Scientific Spectra Multicolor Broad Range Protein Ladder is a 4-color (blue, orange, green, pink) protein standard containing 10 prestained proteins (10 to 260 kDa) for use in gel electrophoresis and western blotting. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.

Applications

  • Monitoring protein migration during SDS-polyacrylamide gel electrophoresis
  • Monitoring protein transfer onto membranes after western blotting
  • Sizing of proteins on SDS-polyacrylamide gels and western blots
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contents: eight vials of 250 μL each

Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3 and 33% glycerol

Storage: Upon receipt store at -20°C

10
Yes
4 colors: Blue, Orange, Green, Pink
260, 140, 100, 70, 50, 40, 35, 25, 15, 10 kDa
8 x 250 μL
SDS-PAGE
Colorimetric, NIR Fluorescence (700 nm), RGB Fluorescence (555 nm)
Protein Ladder
10 to 260 kDa
Bolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Spectra™
Approved for shipment on Wet or Dry Ice
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only. Not for use in diagnostic procedures.