Learn More
Thermo Scientific™ Spectra™ MRSA Medium
Obtain fast and accurate results with a selective chromogenic medium for screening clinical samples for the presence of Methicillin Resistant Staphylococcus aureus.
$128.00 - $1262.00
Specifications
Color | Opaque light tan |
---|---|
Content And Storage | 2°C to 8°C |
Description | Spectra MRSA Medium |
Format | 85mm Monoplate |
Manufacturing Origin | United States |
Catalog Number | Mfr. No. | Description | dimFormat | Quantity | Manufacturing Origin | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | dimFormat | Quantity | Manufacturing Origin | Price | Quantity & Availability | ||
R01821
|
Thermo Scientific™
R01821 |
Spectra MRSA Medium | 10/Pk. | United States |
Pack of 10 for $128.00
|
|
|||
R01822
|
Thermo Scientific™
R01822 |
Spectra MRSA Medium | 100/Pk. | United States |
Case of 100 for $1,262.00
|
|
|||
Description
Obtain faster results with no need for reincubation of negatives with Thermo Scientific™ Spectra™ MRSA, a new, improved chromogenic screening plate for Methicillin Resistant Staphylococcus aureus (MRSA). The medium combines excellent sensitivity with the simplicity of a chromogenic plate to create an invaluable tool in the implementation of infection control policies and procedures.
Limit the spread of MRSA by incorporating an effective screening protocol as part of your infection control program. Colonized patients must be targeted for isolation and appropriate treatment as early as possible; therefore, speed and accuracy of results are recognized as critical. Save time and infection control resources by giving negative patients the all-clear faster and more reliably.- Rapidly and economically detects nasal colonization of MRSA and MRSA from positive blood cultures
- Aids in the prevention of MRSA infections
- Delivers high Positive Predictive Value (PPV) to reduce unnecessary costs
- Enables quick, efficient screening of patients
- Produces easy-to-read, denim blue colonies on an opaque background
- Requires no special handling for optimal workflow 24/7
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Opaque light tan | |
Spectra MRSA Medium | |
United States | |
18 mL |
2°C to 8°C | |
85mm Monoplate | |
Agar | |
Flow Wrapped - Single Layer |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.