Learn More
Perkin Elmer US LLC Spare Standard Mirror for the Relative Specular Reflectance Accessory
Supplier: Perkin Elmer US LLC B0071519
The Spare Standard Mirror is an uncalibrated, flat mirror, approximately 40 mm square. It is used with the Relative Specular Reflectance Accessory 6° (B0086703) and the Variable Angle Reflectance Accessory (B0137314).