Learn More
Bel-Art™ SP Scienceware™ Large Pails
Rugged pails with fitted lids/covers and steel-plated bail handles
Supplier: Bel-Art™ F167760000/EMD
Description
- Available in yellow or translucent white for easier viewing of fluid level
- Polypropylene is steam autoclavable (121°C), Polyethylene is not autoclavable
- 14-qt./13.2L pail has steel-plated swinging bail handle with grip
- Large pails meet US Specification MIL-P36257A
Compliance
Meet US Specification MIL-P36257A (8/10/70).
Specifications
US, MIL-PL36257A | |
White |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov