Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bel-Art™ SP Scienceware™ Biohazard Disposal Bags with/without Warning Label

Economical, safe disposal of used pipettes, Petri dishes, multi-well cell culture plates, cell culture flasks, culture plates, membrane filters and other biohazard wastes
$87.00 - $234.00
Specifications
Certifications/Compliance | ASTM D1709-98 |
---|---|
Type | Biohazard Bag |
Material | Polypropylene |
Disposable | Yes |
Autoclavable | Autoclavable |
Catalog Number | Mfr. No. | Color | Design | Length (English) | Width (English) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Design | Length (English) | Width (English) | Price | Quantity & Availability | ||
S43258
![]() |
Bel-Art™
F131600009/EMD |
Clear | Biohazard Symbol with Sterilization Patch | 12 in. | 24 in. |
Pack of 100 for $89.00
|
|
S43259
![]() |
Bel-Art™
F131610009/EMD |
Clear | Biohazard Symbol with Sterilization Patch | 24 in. | 30 in. |
Pack of 100 for $222.00
|
|
S43261
![]() |
Bel-Art™
F131600005/EMD |
Translucent | Non-printed without Sterilization Patch | 24 in. | 12 in. |
Pack of 100 for $87.00
|
|
S43263
![]() |
Bel-Art™
F131610005/EMD |
Translucent | Non-printed without Sterilization Patch | 30 in. | 24 in. |
Pack of 100 for $196.00
|
|
S43264
![]() |
Bel-Art™
F131620005/EMD |
Translucent | Non-printed without Sterilization Patch | 36 in. | 24 in. |
Pack of 100 for $230.00
|
|
S43268
![]() |
Bel-Art™
F131620009/EMD |
Clear | Biohazard Symbol with Sterilization Patch | 24 in. | 36 in. |
Pack of 100 for $234.00
|
|
Description
- These transparent, polypropylene plastic biohazard bags are 0.03mm (1.5 mil) thick
- Pass ASTM 1709 Dart Impact and ASTM 1922 Tear Resistance Tests
- High temperature autoclavable at a maximum of 135°C (275°F)
- May also be used as conventional trash can liners
Specifications
ASTM D1709-98 | |
Polypropylene | |
Autoclavable | |
1.5 mm |
Biohazard Bag | |
Yes | |
Disposal of used pipettes, petri dishes, culture plates, membrane filters and more |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title