Learn More
Bel-Art™ SP Scienceware™ Battery-Powered Engraving Tool and Stand
Permanently mark important tools, samples and more in seconds
$44.50
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S42963
|
Bel-Art™
F441500005/EMD |
Battery-powered Engraving Tool |
Each for $44.50
|
|
|||||
S42964
|
Bel-Art™
F441500015/EMD |
Battery-powered Engraving Tool Stand | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Engraving Tool (Mfr. No. F441500005)
- Hold the Engraving Tool like a pen, press the power button, apply light pressure, and “write” to permanently mark most materials including glass, plastic, wood, metal, bone, and stone
- Portable, cordless battery power: operates on two “AA” batteries (not included)
- Use indoors or out in the field
- Marks will not fade if exposed to chemicals or sunlight
Engraving Tool Stand (Mfr. No. F441500015)
- Keeps tool handy when not in use
- Molded from polyethylene
- Well to hold extra engraving tip
Order Info
7 in. (17.8cm) long
Specifications
Extra engraving tip |