Learn More
Bel-Art™ SP Scienceware™ Wilmad-LabGlass™ 5mm Spinner Turbine, Var., NMR
High quality 5 mm NMR spinner turbine.
Supplier:  Bel-Art™ 660000051
Catalog No. 16-800-151
Add to Cart
Alert:
Due to the nature of glass, Extreme Caution should be exercised when using at elevated or reduced pressures as performance of tubes under pressure or vacuum cannot be guaranteed. Adequate safety shielding should always be used when working in these conditions.
Description
Description
Specifications
Specifications
Spinner Turbine
5.0 mm