Learn More
Bioworld SOYBEAN AGGLUTININ-LECTIN
Supplier: Bioworld 215100102
Glycine max (soybean) Lectin isolated from Glycine max (soybean) seeds. Specifications: Primary sugar specificity: Galactose/N-acetyl galactosamine Preferential inding activity: terminal alpha or beta-linked N-acetylgalactosamine.