Learn More
NEOGEN Soy Peptone, 500 g
SDP
Supplier:  NEOGEN 700003589
Encompass_Preferred
Soy Peptone is for use in preparing microbiological culture media in a laboratory setting, and is not intended for use in the diagnosis of disease or other conditions in humans. Prepared using the enzyme papain to digest soyabean meal, this peptone is a rich source of nutrients with a high carbohydrate content. Most organisms will grow rapidly in this peptone but some bacteria will produce high levels of acid leading to auto-sterilization unless an adequate buffering system is incorporated.For more information visit https://www.neogen.com/solutions/Microbiology/soy-peptone
Catalog No. 50-201-5816
Explore available promotions